Anti-ITGA9 Mouse Monoclonal Antibody [clone: 560201]

Supplier: R & D Systems
MAB4574
220020-936EA 577.02 USD
220020-936
Anti-ITGA9 Mouse Monoclonal Antibody [clone: 560201]
Antibodies
Type: Primary
Antigen: ITGA9
Clonality: Monoclonal
Clone: 560201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR