Anti-CCL21A Rat Monoclonal Antibody [clone: 709420]

Supplier: R & D Systems
MAB4571
102885-768EA 561.11 USD
102885-768
Anti-CCL21A Rat Monoclonal Antibody [clone: 709420]
Antibodies
Type: Primary
Antigen: CCL21A
Clonality: Monoclonal
Clone: 709420
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR