Anti-S100 A8 Mouse Monoclonal Antibody [clone: 749916]

Supplier: R & D Systems
MAB4570
102885-766EA 600.15 USD
102885-766
Anti-S100 A8 Mouse Monoclonal Antibody [clone: 749916]
Antibodies
Type: Primary
Antigen: S100 A8
Clonality: Monoclonal
Clone: 749916
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR