Anti-ARRB1 Rat Monoclonal Antibody [clone: 425817]

Supplier: R & D Systems
MAB45601
220020-934EA 577.02 USD
220020-934
Anti-ARRB1 Rat Monoclonal Antibody [clone: 425817]
Antibodies
Type: Primary
Antigen: ARRB1
Clonality: Monoclonal
Clone: 425817
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR