Anti-ILKAP Mouse Monoclonal Antibody [clone: 499526]

Supplier: R & D Systems
MAB4554
220020-933EA 561.11 USD
220020-933
Anti-ILKAP Mouse Monoclonal Antibody [clone: 499526]
Antibodies
Type: Primary
Antigen: ILKAP
Clonality: Monoclonal
Clone: 499526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR