Anti-KITLG Rat Monoclonal Antibody [clone: 40215]

Supplier: R & D Systems
MAB455
220020-932EA 772.25 USD
220020-932
Anti-KITLG Rat Monoclonal Antibody [clone: 40215]
Antibodies
Type: Primary
Antigen: KITLG
Clonality: Monoclonal
Clone: 40215
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR