Anti-SIRPA Mouse Monoclonal Antibody [clone: 602411]

Supplier: R & D Systems
MAB4546
102885-764EA 655.11 USD
102885-764
Anti-SIRPA Mouse Monoclonal Antibody [clone: 602411]
Antibodies
Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 602411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR