Anti-GRK5 Mouse Monoclonal Antibody [clone: 691816]

Supplier: R & D Systems
MAB4539
102885-762EA 561.11 USD
102885-762
Anti-GRK5 Mouse Monoclonal Antibody [clone: 691816]
Antibodies
Type: Primary
Antigen Symbol: GRK5
Antigen Name: G protein-coupled receptor kinase 5
Antigen Synonyms: GPRK5
Clonality: Monoclonal
Clone: 691816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR