Anti-FOXP1 Mouse Monoclonal Antibody [clone: 542721]

Supplier: R & D Systems
MAB4534
220020-930EA 561.11 USD
220020-930
Anti-FOXP1 Mouse Monoclonal Antibody [clone: 542721]
Antibodies
Type: Primary
Antigen Symbol: FOXP1
Antigen Name: forkhead box P1
Antigen Synonyms: 12CC4, HSPC215, MFH, QRF1, hFKH1B
Clonality: Monoclonal
Clone: 542721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR