Anti-PTK2 Rat Monoclonal Antibody [clone: 820755]

Supplier: R & D Systems
MAB4528
102885-758EA 594.37 USD
102885-758
Anti-PTK2 Rat Monoclonal Antibody [clone: 820755]
Antibodies
Type: Primary
Antigen: PTK2
Clonality: Monoclonal
Clone: 820755
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR