Anti-ELANE Rat Monoclonal Antibody [clone: 887105]

Supplier: R & D Systems
MAB4517
102885-756EA 616.06 USD
102885-756
Anti-ELANE Rat Monoclonal Antibody [clone: 887105]
Antibodies
Type: Primary
Antigen: ELANE
Clonality: Monoclonal
Clone: 887105
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR