Anti-CDCP1 Rat Monoclonal Antibody [clone: 469114]

Supplier: R & D Systems
MAB45151
220020-925EA 594.37 USD
220020-925
Anti-CDCP1 Rat Monoclonal Antibody [clone: 469114]
Antibodies
Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 469114
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR