Anti-TSHB Mouse Monoclonal Antibody [clone: 442506]

Supplier: R & D Systems
MAB4507
220020-921EA 616.06 USD
220020-921
Anti-TSHB Mouse Monoclonal Antibody [clone: 442506]
Antibodies
Type: Primary
Antigen: TSHB
Clonality: Monoclonal
Clone: 442506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR