Anti-EMP3 Mouse Monoclonal Antibody [clone: 439949]

Supplier: R & D Systems
MAB4505
220020-920EA 577.02 USD
220020-920
Anti-EMP3 Mouse Monoclonal Antibody [clone: 439949]
Antibodies
Type: Primary
Antigen: EMP3
Clonality: Monoclonal
Clone: 439949
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR