Anti-MS4A4B Rat Monoclonal Antibody [clone: 444008]

Supplier: R & D Systems
MAB4504
220020-919EA 577.02 USD
220020-919
Anti-MS4A4B Rat Monoclonal Antibody [clone: 444008]
Antibodies
Type: Primary
Antigen: MS4A4B
Clonality: Monoclonal
Clone: 444008
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR