Anti-CCL3 Rat Monoclonal Antibody [clone: 756613]

Supplier: R & D Systems
MAB4501
102885-754EA 594.37 USD
102885-754
Anti-CCL3 Rat Monoclonal Antibody [clone: 756613]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 756613
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR