Anti-PTK7 Mouse Monoclonal Antibody [clone: 525222]

Supplier: R & D Systems
MAB4499
220020-916EA 600.15 USD
220020-916
Anti-PTK7 Mouse Monoclonal Antibody [clone: 525222]
Antibodies
Simple Western

Type: Primary
Antigen: PTK7
Clonality: Monoclonal
Clone: 525222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR