Anti-GRK1 Mouse Monoclonal Antibody [clone: 466630]

Supplier: R & D Systems
MAB4498
220020-915EA 561.11 USD
220020-915
Anti-GRK1 Mouse Monoclonal Antibody [clone: 466630]
Antibodies
Type: Primary
Antigen Symbol: GRK1
Antigen Name: G protein-coupled receptor kinase 1
Antigen Synonyms: GPRK1, RHOK, RK
Clonality: Monoclonal
Clone: 466630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR