Anti-EDNRB Mouse Monoclonal Antibody [clone: 671917]

Supplier: R & D Systems
MAB4496
102885-752EA 594.37 USD
102885-752
Anti-EDNRB Mouse Monoclonal Antibody [clone: 671917]
Antibodies
Type: Primary
Antigen: EDNRB
Clonality: Monoclonal
Clone: 671917
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR