Anti-NAAA Mouse Monoclonal Antibody [clone: 511702]

Supplier: R & D Systems
MAB4494
220020-914EA 584.25 USD
220020-914
Anti-NAAA Mouse Monoclonal Antibody [clone: 511702]
Antibodies
Type: Primary
Antigen: NAAA
Clonality: Monoclonal
Clone: 511702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR