Anti-HPX Mouse Monoclonal Antibody [clone: 698813]

Supplier: R & D Systems
MAB4490
102885-750EA 577.02 USD
102885-750
Anti-HPX Mouse Monoclonal Antibody [clone: 698813]
Antibodies
Type: Primary
Antigen: HPX
Clonality: Monoclonal
Clone: 698813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR