Anti-LIF Rat Monoclonal Antibody [clone: 139124]

Supplier: R & D Systems
MAB449
220020-913EA 763.57 USD
220020-913
Anti-LIF Rat Monoclonal Antibody [clone: 139124]
Antibodies
Type: Primary
Antigen: LIF
Clonality: Monoclonal
Clone: 139124
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR