Anti-BoNT-A Mouse Monoclonal Antibody [clone: 503013]

Supplier: R & D Systems
MAB4489
220020-912EA 600.15 USD
220020-912
Anti-BoNT-A Mouse Monoclonal Antibody [clone: 503013]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: BoNT-A Light Chain
Antigen Synonyms:
Clonality: Monoclonal
Clone: 503013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: C. botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR