Anti-SIRPG Mouse Monoclonal Antibody [clone: 595337]

Supplier: R & D Systems
MAB4486
102885-748EA 594.37 USD
102885-748
Anti-SIRPG Mouse Monoclonal Antibody [clone: 595337]
Antibodies
Type: Primary
Antigen: SIRPG
Clonality: Monoclonal
Clone: 595337
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR