Anti-IL17RA Rat Monoclonal Antibody [clone: 105828]

Supplier: R & D Systems
MAB448
220020-909EA 763.57 USD
220020-909
Anti-IL17RA Rat Monoclonal Antibody [clone: 105828]
Antibodies
Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 105828
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR