Anti-SLURP1 Mouse Monoclonal Antibody [clone: 569317]

Supplier: R & D Systems
MAB4479
102885-744EA 566.89 USD
102885-744
Anti-SLURP1 Mouse Monoclonal Antibody [clone: 569317]
Antibodies
Type: Primary
Antigen: SLURP1
Clonality: Monoclonal
Clone: 569317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR