Anti-S100 A7 Mouse Monoclonal Antibody [clone: 577513]

Supplier: R & D Systems
MAB4475
102885-742EA 577.02 USD
102885-742
Anti-S100 A7 Mouse Monoclonal Antibody [clone: 577513]
Antibodies
Type: Primary
Antigen: S100 A7
Clonality: Monoclonal
Clone: 577513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR