Anti-MYHC Mouse Monoclonal Antibody [clone: MF20]

Supplier: R & D Systems
MAB4470
220020-908EA 639.2 USD
220020-908
Anti-MYHC Mouse Monoclonal Antibody [clone: MF20]
Antibodies
Type: Primary
Antigen: MYHC
Clonality: Monoclonal
Clone: MF20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR