Anti-PTK2 Mouse Monoclonal Antibody [clone: 495919]

Supplier: R & D Systems
MAB4467
220020-906EA 405.66 USD
220020-906
Anti-PTK2 Mouse Monoclonal Antibody [clone: 495919]
Antibodies
Type: Primary
Antigen: PTK2
Clonality: Monoclonal
Clone: 495919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR