Anti-PRKCI+PRKCL Mouse Monoclonal Antibody [clone: 450401]

Supplier: R & D Systems
MAB4465
220020-905EA 577.02 USD
220020-905
Anti-PRKCI+PRKCL Mouse Monoclonal Antibody [clone: 450401]
Antibodies
Type: Primary
Antigen: PRKCI+PRKCL
Clonality: Monoclonal
Clone: 450401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR