Anti-EPHB4 Rat Monoclonal Antibody [clone: 117808]

Supplier: R & D Systems
MAB446
220020-904EA 772.25 USD
220020-904
Anti-EPHB4 Rat Monoclonal Antibody [clone: 117808]
Antibodies
Type: Primary
Antigen: EPHB4
Clonality: Monoclonal
Clone: 117808
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR