Anti-JUNB Mouse Monoclonal Antibody [clone: 512313]

Supplier: R & D Systems
MAB4456
102885-736EA 577.02 USD
102885-736
Anti-JUNB Mouse Monoclonal Antibody [clone: 512313]
Antibodies
Type: Primary
Antigen: JUNB
Clonality: Monoclonal
Clone: 512313
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR