Anti-SHH Rat Monoclonal Antibody [clone: 55626]

Supplier: R & D Systems
MAB445
220020-899EA 763.57 USD
220020-899
Anti-SHH Rat Monoclonal Antibody [clone: 55626]
Antibodies
Type: Primary
Antigen: SHH
Clonality: Monoclonal
Clone: 55626
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR