Anti-SSTR5 Mouse Monoclonal Antibody [clone: 394401]

Supplier: R & D Systems
MAB4448
220020-897EA 594.37 USD
220020-897
Anti-SSTR5 Mouse Monoclonal Antibody [clone: 394401]
Antibodies
Type: Primary
Antigen: SSTR5
Clonality: Monoclonal
Clone: 394401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR