Anti-REN1 Mouse Monoclonal Antibody [clone: 411507]

Supplier: R & D Systems
MAB4447
220020-896EA 594.37 USD
220020-896
Anti-REN1 Mouse Monoclonal Antibody [clone: 411507]
Antibodies
Type: Primary
Antigen: REN1
Clonality: Monoclonal
Clone: 411507
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR