Anti-PLOD2 Mouse Monoclonal Antibody [clone: 408105]

Supplier: R & D Systems
MAB4445
220020-894EA 647.88 USD
220020-894
Anti-PLOD2 Mouse Monoclonal Antibody [clone: 408105]
Antibodies
Type: Primary
Antigen: PLOD2
Clonality: Monoclonal
Clone: 408105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR