Anti-CD226 Rat Monoclonal Antibody [clone: 838216]

Supplier: R & D Systems
MAB4436
102885-734EA 577.02 USD
102885-734
Anti-CD226 Rat Monoclonal Antibody [clone: 838216]
Antibodies
Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: 838216
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR