Anti-KDR Rat Monoclonal Antibody [clone: 522302]

Supplier: R & D Systems
MAB4432
220020-893EA 594.37 USD
220020-893
Anti-KDR Rat Monoclonal Antibody [clone: 522302]
Antibodies
Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: 522302
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR