Anti-KDR Rat Monoclonal Antibody [clone: 91201]

Supplier: R & D Systems
MAB443
220020-890EA 772.25 USD
220020-890
Anti-KDR Rat Monoclonal Antibody [clone: 91201]
Antibodies
Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: 91201
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR