Anti-ADAM12 Mouse Monoclonal Antibody [clone: 632525]

Supplier: R & D Systems
MAB44161
102885-732EA 600.15 USD
102885-732
Anti-ADAM12 Mouse Monoclonal Antibody [clone: 632525]
Antibodies
Type: Primary
Antigen: ADAM12
Clonality: Monoclonal
Clone: 632525
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR