Anti-ARSB Mouse Monoclonal Antibody [clone: 708401]

Supplier: R & D Systems
MAB4415
102885-730EA 600.15 USD
102885-730
Anti-ARSB Mouse Monoclonal Antibody [clone: 708401]
Antibodies
Type: Primary
Antigen: ARSB
Clonality: Monoclonal
Clone: 708401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR