Anti-SERPINA12 Mouse Monoclonal Antibody [clone: 484223]

Supplier: R & D Systems
MAB4410
220020-888EA 561.11 USD
220020-888
Anti-SERPINA12 Mouse Monoclonal Antibody [clone: 484223]
Antibodies
Type: Primary
Antigen: SERPINA12
Clonality: Monoclonal
Clone: 484223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR