Anti-TFF3 Mouse Monoclonal Antibody [clone: 415921]

Supplier: R & D Systems
MAB4408
220020-887EA 639.2 USD
220020-887
Anti-TFF3 Mouse Monoclonal Antibody [clone: 415921]
Antibodies
Type: Primary
Antigen: TFF3
Clonality: Monoclonal
Clone: 415921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR