Anti-TCF3 Rat Monoclonal Antibody [clone: 401709]

Supplier: R & D Systems
MAB44061
220020-885EA 561.11 USD
220020-885
Anti-TCF3 Rat Monoclonal Antibody [clone: 401709]
Antibodies
Type: Primary
Antigen: TCF3
Clonality: Monoclonal
Clone: 401709
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR