Anti-GRM7 Mouse Monoclonal Antibody [clone: 437310]

Supplier: R & D Systems
MAB4405
220020-884EA 577.02 USD
220020-884
Anti-GRM7 Mouse Monoclonal Antibody [clone: 437310]
Antibodies
Type: Primary
Antigen Symbol: GRM7
Antigen Name: glutamate receptor, metabotropic 7
Antigen Synonyms: GLUR7, GPRC1G, MGLU7, MGLUR7, PPP1R87
Clonality: Monoclonal
Clone: 437310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR