Anti-SCD Rat Monoclonal Antibody [clone: 428208]

Supplier: R & D Systems
MAB4404
220020-883EA 594.37 USD
220020-883
Anti-SCD Rat Monoclonal Antibody [clone: 428208]
Antibodies
Type: Primary
Antigen: SCD
Clonality: Monoclonal
Clone: 428208
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR