Anti-CD40 Rat Monoclonal Antibody [clone: 65924]

Supplier: R & D Systems
MAB4401
220020-882EA 772.25 USD
220020-882
Anti-CD40 Rat Monoclonal Antibody [clone: 65924]
Antibodies
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 65924
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR