Anti-ENTHD1 Rat Monoclonal Antibody [clone: 495826]

Supplier: R & D Systems
MAB4398
220020-879EA 647.88 USD
220020-879
Anti-ENTHD1 Rat Monoclonal Antibody [clone: 495826]
Antibodies
Type: Primary
Antigen: ENTHD1
Clonality: Monoclonal
Clone: 495826
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR