Anti-ENTHD1 Mouse Monoclonal Antibody [clone: 498409]

Supplier: R & D Systems
MAB43971
220020-878EA 594.37 USD
220020-878
Anti-ENTHD1 Mouse Monoclonal Antibody [clone: 498409]
Antibodies
Type: Primary
Antigen: ENTHD1
Clonality: Monoclonal
Clone: 498409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR