Anti-CCL22 Rat Monoclonal Antibody [clone: 158113]

Supplier: R & D Systems
MAB439
220020-875EA 763.57 USD
220020-875
Anti-CCL22 Rat Monoclonal Antibody [clone: 158113]
Antibodies
Type: Primary
Antigen: CCL22
Clonality: Monoclonal
Clone: 158113
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR